Order Entry
Netherlands
ContactUsLinkComponent
2-Chloro-3-(chloromethyl)-7-methoxyquinoline
2-Chloro-3-(chloromethyl)-7-methoxyquinoline
Catalog # APOSOR307909-1G
Supplier:  Apollo Scientific
CAS Number:  
2-Chloro-3-(chloromethyl)-7-methoxyquinoline
Catalog # APOSOR307909-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Storage Temperature:Ambient
CAS Number:73863-49-7
Formula:C₁₁H₉Cl₂NO