Order Entry
Netherlands
ContactUsLinkComponent
(R)-1-Boc-piperidine-3-carboxylic acid 95+%
(R)-1-Boc-piperidine-3-carboxylic acid 95+%
Catalog # APOSOR307102-25G
Supplier:  Apollo Scientific
CAS Number:  
(R)-1-Boc-piperidine-3-carboxylic acid 95+%
Catalog # APOSOR307102-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :229.28 g/mol
Storage Temperature:Ambient
CAS Number:163438-09-3
Formula:C₁₁H₁₉NO₄
MDL Number:MFCD02179173