
(R)-(+)-2-Amino-1,1-diphenyl-1-propanol
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:78603-93-7
:
Formula:C₁₅H₁₇NO
MDL Number:MFCD02677967
:227.30 g/mol
Melting Pt:102…105 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together