6-[(4-Methoxyphenyl)sulphonyl]-3-pyridinylamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1221792-06-8
:
Formula:C₁₂H₁₂N₂O₃S
MDL Number:MFCD16140353
:264.31 g/mol
Melting Pt:180…182 °C
Storage Temperature:Ambient
Frequently Bought Together