tert-Butyl-4-(methoxymethyl)piperidine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:509147-78-8
:
Formula:C₁₂H₂₃NO₃
MDL Number:MFCD07779470
:229.32 g/mol
Storage Temperature:Ambient
Frequently Bought Together