1-[(4-Isothiocyanatophenyl)sulphonyl]piperidine
Catalog # APOSOR30283-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:282.39 g/mol
Storage Temperature:Ambient
CAS Number:7356-55-0
Formula:C₁₂H₁₄N₂O₂S₂
MDL Number:MFCD00023666