
tert-Butyl-4-(bromomethyl)piperidine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:158407-04-6
:
Formula:C₁₁H₂₀BrNO₂
MDL Number:MFCD04115538
:278.1860 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together