Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(2-Pyrrolidin-1-ylethoxy)phenylamine
4-(2-Pyrrolidin-1-ylethoxy)phenylamine
4-(2-Pyrrolidin-1-ylethoxy)phenylamine
APOSOR302272-1G
:
Apollo Scientific
:
4-(2-Pyrrolidin-1-ylethoxy)phenylamine
APOSOR302272-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
206.29 g/mol
:
Ambient
:
50609-01-3
:
C₁₂H₁₈N₂O
:
MFCD07790198