1-(2-Tetrahydropyranyl)-1H-pyrazole 97%
:449758-17-2
:
:C₈H₁₂N₂O
:MFCD11044692
:152.20 g/mol
:60 °C
:104.4 °C
:Ambient
Frequently Bought Together
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
n-Pentane ≥99%, GPR RECTAPUR®
: VWR Chemicals
Ethyl acetate 99.6% ACS, meets analytical specifications of Reagent Ph. Eur.
: Thermo Fisher Scientific
n-Heptane ∼ 99% (n-heptane) for HPLC
: Thermo Fisher Scientific
N,N-Dimethylformamide, anhydrous ≥99.8%
: Thermo Fisher Scientific
1-(2-Tetrahydropyranyl)-1H-pyrazole-5-boronic acid pinacol ester
: Apollo Scientific
Benzonitrile 99+% for spectroscopy
: Thermo Fisher Scientific
2-Methylbutane ≥99%, Reagent Grade
: Honeywell Chemicals
Acetonitrile, extra dry over Molecular Sieve 99.9%, AcroSeal®
: Thermo Fisher Scientific
Petroleum spirit, 80…110 °C
: Roth Carl