1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:864754-16-5
:
Formula:C₁₃H₂₁BN₂O₄
MDL Number:MFCD08706026
:280.1278 g/mol
Storage Temperature:Ambient
Frequently Bought Together