Order Entry
Netherlands
ContactUsLinkComponent
7-Hydroxy-1-indanone
Irritant
7-Hydroxy-1-indanone
Catalog # APOSOR301315-5G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
7-Hydroxy-1-indanone
Catalog # APOSOR301315-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
Boiling Pt:131 °C/9.5mmHg
Melting Pt:109…113 °C
 :148.161 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:6968-35-0
Formula:C₉H₈O₂
MDL Number:MFCD01548387