Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-2-methoxybenzonitrile
5-Chloro-2-methoxybenzonitrile
5-Chloro-2-methoxybenzonitrile
5-Chloro-2-methoxybenzonitrile
APOSOR30129-250MG
:
Apollo Scientific
:
5-Chloro-2-methoxybenzonitrile
APOSOR30129-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
Environmentally Preferable:
:
:
167.59 g/mol
:
3439
:
GHS06
:
Ambient
:
55877-79-7
:
C₈H₆ClNO
:
MFCD00052867
:
276
:
98…102 °C
:
6.1 III