Order Entry
Netherlands
ContactUsLinkComponent
2-Isopropylphenyl isothiocyanate
Corrosive
Irritant
2-Isopropylphenyl isothiocyanate
Catalog # APOSOR28865-25G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
2-Isopropylphenyl isothiocyanate
Catalog # APOSOR28865-25G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :177.27 g/mol
Boiling Pt:131…132 °C/10mmHg
Density:1.05 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2922
ADR:8+6.1,III
 :GHS05
 :GHS07
CAS Number:36176-31-5
Formula:C₁₀H₁₁NS
MDL Number:MFCD00022053