
Ethyl-5-bromo-2-chlorobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:76008-73-6
:
Formula:C₉H₈BrClO₂
MDL Number:MFCD00013635
:263.52 g/mol
Boiling Pt:288 °C
Density:1.55 g/cm³ (25 °C)
Flash Pt:>110 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together