


2-Chloro-6-(trichloromethyl)pyridine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1929-82-4
:
Formula:C₆H₃Cl₄N
MDL Number:MFCD00072500
:230.897 g/mol
Boiling Pt:136…138 °C/11mmHg
Melting Pt:63…65 °C
Flash Pt:110 °C
EINECS:217-682-2
UN:2811
ADR:6.1,III
:GHS06
:GHS07
:GHS09
Storage Temperature:Ambient
Frequently Bought Together