

6-Methyl-2-thiouracil
Catalog # APOSOR27585-50G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:50 g
- Seq:3368
Specifications
About this item
:
:142.18 g/mol
Melting Pt:330 °C (dec.)
Density:1.36 g/cm³ (25 °C)
EINECS:200-252-3
:GHS07
:GHS08
Storage Temperature:Ambient
CAS Number:56-04-2
:MFCD00006040
Formula:C₅H₆N₂OS