Order Entry
Netherlands
ContactUsLinkComponent
4-Hydrazino-7-methylthieno[3,2-d]pyrimidine
4-Hydrazino-7-methylthieno[3,2-d]pyrimidine
Catalog # APOSOR24332-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Hydrazino-7-methylthieno[3,2-d]pyrimidine
Catalog # APOSOR24332-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :180.23 g/mol
Storage Temperature:Ambient
CAS Number:175137-22-1
Formula:C₇H₈N₄S
MDL Number:MFCD00205196