
(Methyl)triphenylphosphonium iodide 97%
Catalog # APOSOR23590-500G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:500 g
- Seq:3645
Specifications
About this item
:
:404.23 g/mol
Melting Pt:185…188 °C
Storage Temperature:Ambient
UN:3464
ADR:6.1,III
:GHS06
CAS Number:2065-66-9
Formula:C₁₉H₁₈IP
MDL Number:MFCD00066175