Order Entry
Netherlands
ContactUsLinkComponent
1,3-Dimethyl-1H-pyrazol-5-carbaldehyde
Irritant
1,3-Dimethyl-1H-pyrazol-5-carbaldehyde
Catalog # APOSOR23187-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1,3-Dimethyl-1H-pyrazol-5-carbaldehyde
Catalog # APOSOR23187-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :124.1400 g/mol
Melting Pt:40…42 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:25016-09-5
Formula:C₆H₈N₂O
MDL Number:MFCD02681907