Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Bestelling invoeren
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Benzo[b]thiophen-2-ylmethanamine
Benzo[b]thiophen-2-ylmethanamine
Benzo[b]thiophen-2-ylmethanamine
Benzo[b]thiophen-2-ylmethanamine
APOSOR23170-250MG
:
Apollo Scientific
:
Benzo[b]thiophen-2-ylmethanamine
APOSOR23170-250MG
:
Apollo Scientific
:
Verpakking:
Bottle
VPE:
250 mg
Seq:
2590
:
:
163.23 g/mol
:
58…59 °C
:
Koelkast
:
3259
:
8,III
:
GHS05
:
6314-43-8
:
MFCD00965305
:
C₉H₉NS