
(R)-2-Aminopropanamide hydrochloride 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:71810-97-4
:
Formula:C₃H₉ClN₂O
MDL Number:MFCD00039093
:124.57 g/mol
Melting Pt:169…172 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together