Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Netherlands
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
[1-(3-Chlorophenyl)pyrrolidin-3-yl]methanol
[1-(3-Chlorophenyl)pyrrolidin-3-yl]methanol
[1-(3-Chlorophenyl)pyrrolidin-3-yl]methanol
APOSOR200134-1G
:
Apollo Scientific
:
[1-(3-Chlorophenyl)pyrrolidin-3-yl]methanol
APOSOR200134-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
211.69 g/mol
:
Ambient
:
1017444-94-8
:
C₁₁H₁₄ClNO
:
MFCD10003937