
tert-Butyl-3-(hydroxymethyl)-3-phenylpyrrolidine-1-carboxylate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1314394-67-6
:
Formula:C₁₆H₂₃NO₃
MDL Number:MFCD18064983
:277.37 g/mol
:GHS07
Storage Temperature:Ambient
Frequently Bought Together