
3-(3-Bromophenyl)-1H-pyrazole
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:149739-65-1
Formula:C₉H₇BrN₂
MDL Number:MFCD01940433
:223.07 g/mol
Melting Pt:73…74 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together