
1,5-Dimethyl-1H-pyrazole-4-boronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:1204333-57-2
:
Formula:C₅H₉BN₂O₂
MDL Number:MFCD13195678
:139.9482 g/mol
:GHS07
Storage Temperature:Freezer
Frequently Bought Together