4-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid (cis and trans mixture) 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:130309-46-5
:
Formula:C₁₂H₂₁NO₄
MDL Number:MFCD03453262
:243.30 g/mol
Storage Temperature:Ambient
Frequently Bought Together