
6-Aminopyridin-3-ol hydrochloride 95%
Catalog # APOSOR16984-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:146.58 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:856965-37-2
Formula:C₅H₇ClN₂O
MDL Number:MFCD11506259