Order Entry
Netherlands
ContactUsLinkComponent
6-Oxa-1-azaspiro[3.3]heptane oxalate
Irritant
6-Oxa-1-azaspiro[3.3]heptane oxalate
Catalog # APOSOR16706-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
6-Oxa-1-azaspiro[3.3]heptane oxalate
Catalog # APOSOR16706-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :189.17 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1359655-43-8
Formula:C₇H₁₁NO₅
MDL Number:MFCD21364487