

Methyl(2-methyl-2H-pyrazol-3-ylmethyl)amine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:930111-04-9
:
Formula:C₆H₁₁N₃
MDL Number:MFCD03419812
:125.17 g/mol
Boiling Pt:102 °C/15mmHg
UN:2922
ADR:8+6.1,III
:GHS05
:GHS07
Storage Temperature:Ambient
Frequently Bought Together