
Methyl-5-bromo-2-iodobenzoate
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:181765-86-6
:
Formula:C₈H₆BrIO₂
MDL Number:MFCD00144771
:340.9400 g/mol
Melting Pt:45…49 °C
Flash Pt:>110 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1,III
:GHS06
Frequently Bought Together