(S)-1-[(9H-Fluoren-9-ylmethoxy)carbonyl]-2-piperidinecarboxylic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:86069-86-5
:
Formula:C₂₁H₂₁NO₄
MDL Number:MFCD00235898
:351.40 g/mol
Storage Temperature:Ambient
Frequently Bought Together