N-(tert-Butoxycarbonyl)-4-nitro-D-phenylalanine 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:61280-75-9
:
Formula:C₁₄H₁₈N₂O₆
MDL Number:MFCD00069880
:310.31 g/mol
Storage Temperature:Ambient
Frequently Bought Together