
2-Hydroxy-2-(2-hydroxy-1-naphthyl)acetic acid
Catalog # APOSOR14276-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
:218.21 g/mol
Melting Pt:129…130 °C
:GHS07
Storage Temperature:Ambient
CAS Number:624722-10-7
Formula:C₁₂H₁₀O₄
MDL Number:MFCD06411618