Order Entry
Netherlands
ContactUsLinkComponent
1-(3-Bromophenyl)-1H-pyrazole 97%
Irritant
1-(3-Bromophenyl)-1H-pyrazole 97%
Catalog # APOSOR1418-1G
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-(3-Bromophenyl)-1H-pyrazole 97%
Catalog # APOSOR1418-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :223.0693 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:294877-33-1
Formula:C₉H₇BrN₂
MDL Number:MFCD06797472