2-[(3,4-Dichlorobenzyl)sulphanyl]-1-methyl-1H-imidazole-5-carbaldehyde
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:924869-11-4
:
Formula:C₁₂H₁₀Cl₂N₂OS
MDL Number:MFCD08689685
:301.2 g/mol
Melting Pt:95…97 °C
Storage Temperature:Ambient
Frequently Bought Together