
6-Chloro-1-benzothiophene-2-carboxylic acid
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:26018-73-5
:
Formula:C₉H₅ClO₂S
MDL Number:MFCD07377059
:212.66 g/mol
Melting Pt:271…274 °C
:GHS07
Storage Temperature:Kamertemperatuur
Frequently Bought Together