Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Morpholine 99%

Morpholine 99%

Corrosive
Flammable
Irritant
Supplier:   Apollo Scientific
Morpholine 99%
Supplier:  Apollo Scientific
Certificates

3 Options Available Below

Product Details & Documents
  • CAS Number:110-91-8
  • Formula:C₄H₉NO
  • MDL Number:MFCD00005972
  • Molecular Weight:87.12 g/mol
  • Boiling Pt:129 °C
  • Melting Pt:–7…–5 °C
  • Melting Pt:–7 to –5 °C
  • Density:0.996 g/cm³ (25 °C)
  • Flash Pt:31 °C
  • Storage Temperature:Ambient
  • EINECS:203-815-1
  • UN:2054
  • ADR:8+3,I
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 29,00 €
    29,00 €
    1 * 100 g
       29,00 €

  • Bottle
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 123,00 €
    123,00 €
    1 * 500 g
       123,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 2,5 kg
    1 * 2,5 kg - 615,00 €
    615,00 €
    1 * 2,5 kg
       615,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.