Order Entry
Netherlands
Orders LinkContactUsLinkComponent
 

Nipecotamide

Irritant
New Product
Leverancier:   Apollo Scientific
Nipecotamide
Leverancier:  Apollo Scientific
New Product
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  Piperidine-3-carboxamide
  • CAS Number:4138-26-5
  • Formula:C₆H₁₂N₂O
  • MDL Number:MFCD00005993
  • Molecular Weight:128.1700 g/mol
  • Melting Pt:103…106 °C
  • Storage Temperature:Ambient
  • EINECS:223-962-5
Product Family Options

Product Information

  • Pack type
    Beschikbaarheid
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 88,30 €
    88,30 €
    1 * 100 g
       88,30 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 372,00 €
    372,00 €
    1 * 500 g
       372,00 €

    Specifications:
    New Product
    New Product