2-(Chloromethyl)-1-methylpiperidine hydrochloride
Catalog # APOSOR11727-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:184.11 g/mol
Storage Temperature:Ambient
CAS Number:27483-92-7
Formula:C₇H₁₅Cl₂N
MDL Number:MFCD09800596