
2-Amino-6-chloropyrazine 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:33332-28-4
:
Formula:C₄H₄ClN₃
MDL Number:MFCD00055024
:129.55 g/mol
Melting Pt:150…152 °C
:GHS07
Storage Temperature:Ambient
Frequently Bought Together
Acetonitrile ≥99.8%, SupraSolv® for GC-MS
Supplier: Merck
Acetonitrile ≥99.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Glycerine ≥99.5%, AnalaR NORMAPUR® ACS analytical reagent, redistilled
Supplier: VWR Chemicals
n-Hexane ≥95%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Ethyl acetate ≥99.7%, PESTINORM® for pesticide residue analysis
Supplier: VWR Chemicals
Tetrahydrofuran ≥99.9% unstabilised, HiPerSolv CHROMANORM® for LC-MS
Supplier: VWR Chemicals
Diisopropyl ether stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
n-Hexane, EMSURE® ACS for analysis, Supelco®
Supplier: Merck
Ethanol absolute ≥99.8%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Acetone ≥99.9%, PESTINORM® Supra Trace for organic trace analysis
Supplier: VWR Chemicals
N-Methyl-2-pyrrolidone (NMP) ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
n-Hexane, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
2-Propanol ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
1-Butylboronic acid, Supelco®
Supplier: Merck
Acetonitrile ≥99.5% (by GC), Puriss. p.a. ACS, Reag. Ph. Eur., Riedel-de Haën™
Supplier: Honeywell Chemicals
Turpentine oil
Supplier: Roth Carl