Order Entry
Netherlands
Orders LinkContactUsLinkComponent
6,7-Dimethoxyisoquinoline 96%

6,7-Dimethoxyisoquinoline 96%

Irritant
Supplier:   Apollo Scientific
6,7-Dimethoxyisoquinoline 96%
Supplier:  Apollo Scientific
Certificates
6,7-Dimethoxyisoquinoline 96%

4 Options Available Below

Product Details & Documents
  • CAS Number:15248-39-2
  • Formula:C₁₁H₁₁NO₂
  • MDL Number:MFCD00666134
  • Boiling Pt:158 °C
  • Melting Pt:94 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Availability
    Price
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg - 34,80 €
    34,80 €
    1 * 100 mg
       34,80 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - 59,40 €
    59,40 €
    1 * 250 mg
       59,40 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 598,00 €
    598,00 €
    1 * 5 g
       598,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 2 870,00 €
    2 870,00 €
    1 * 25 g
       2 870,00 €

    Specifications:
    New Product
    New Product
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.