Order Entry
Netherlands
ContactUsLinkComponent
Methyl (7-methoxy-1-naphthyl)acetate
Methyl (7-methoxy-1-naphthyl)acetate
Catalog # APOSOR111472-250MG
Supplier:  Apollo Scientific
CAS Number:  
Methyl (7-methoxy-1-naphthyl)acetate
Catalog # APOSOR111472-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :230.27 g/mol
Storage Temperature:Ambient
CAS Number:185336-03-2
Formula:C₁₄H₁₄O₃
MDL Number:MFCD18413138