4-[3-Nitro-4-(piperidin-1-yl)phenyl]-1,2-dihydrophthalazin-1-one
Catalog # APOSOR110185-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:350.38 g/mol
Storage Temperature:Ambient
CAS Number:218144-45-7
Formula:C₁₉H₁₈N₄O₃
MDL Number:MFCD00116203