Order Entry
Netherlands
ContactUsLinkComponent
1-Piperidinyl(2-piperidinyl)methanone hydrochloride
1-Piperidinyl(2-piperidinyl)methanone hydrochloride
Catalog # APOSOR110025-1G
Supplier:  Apollo Scientific
CAS Number:  
1-Piperidinyl(2-piperidinyl)methanone hydrochloride
Catalog # APOSOR110025-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :232.76 g/mol
Storage Temperature:Ambient
CAS Number:130497-28-8
Formula:C₁₁H₂₁ClN₂O
MDL Number:MFCD06407996