
3-Hydroxy-4-methyl-2(3H)-thiazolethione
Catalog # APOSOR10884-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:147.21 g/mol
Melting Pt:92…94 °C
:GHS07
Storage Temperature:Ambient
CAS Number:49762-08-5
Formula:C₄H₅NOS₂
MDL Number:MFCD00077793