

Zinc(II) nitrate hydrate 99.99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:13778-30-8
:
Formula:N₂O₆Zn·H₂O
MDL Number:MFCD00149888
:207.42 g/mol
Melting Pt:36 °C
Density:2.065 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-943-8
UN:1514
ADR:5.1,II
:GHS03
:GHS07
Frequently Bought Together