Order Entry
Netherlands
ContactUsLinkComponent
Saponin (from Quillaja Bark)
Saponin (from Quillaja Bark)
Catalog # APOSBIS4332-1KG
Supplier:  Apollo Scientific
CAS Number:  
Saponin (from Quillaja Bark)
Catalog # APOSBIS4332-1KG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
Melting Pt:158 °C (dec.)
Density:0.5 g/cm³ (25 °C)
EINECS:232-462-6
 :GHS07
Storage Temperature:Ambient
CAS Number:8047-15-2
MDL Number:MFCD00081981