Bis-PEG4-acid
Catalog # APOSBIPG1249-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4,7,10,13-Tetraoxahexadecane-1,16-dioic acid
:
:294.30 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:31127-85-2
Formula:C₁₂H₂₂O₈
MDL Number:MFCD22201545