Order Entry
Netherlands
Orders LinkContactUsLinkComponent
Melatonin

Melatonin

Bottle, 1 g
Catalog # APOSBIM0105-1G
Irritant
Supplier:   Apollo Scientific
Melatonin
Catalog # APOSBIM0105-1G
Supplier:  Apollo Scientific
CAS Number:  73-31-4
Melatonin

Bottle, 1 g

CAS Number:  73-31-4
Price
21,80 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  Melatonin, N-(2-(5-Methoxy-1H-indol-3-yl)ethyl)acetamide
  • CAS Number:73-31-4
  • Formula:C₁₃H₁₆N₂O₂
  • MDL Number:MFCD00005655
  • Molecular Weight:232.28 g/mol
  • Melting Pt:116…118 °C
  • Storage Temperature:Freezer
  • EINECS:200-797-7
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.