Order Entry
Netherlands
ContactUsLinkComponent
Fasudil dihydrochloride ≥99%
Irritant
Fasudil dihydrochloride ≥99%
Catalogus # APOSBIH1037-10MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
Fasudil dihydrochloride ≥99%
Catalogus # APOSBIH1037-10MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :364.29 g/mol
Melting Pt:248…249 °C
 :GHS07
Storage Temperature:Freezer
CAS Number:203911-27-7
 :103745-39-7
Formula:C₁₄H₁₇N₃O₂S·2HCl
MDL Number:MFCD00153805